Learn More
Supertek™ Economy Chromatic Set of 13 Tuning Forks
GREENER_CHOICESCI_ED
Catalog No. S81831 Shop All Supertek Scientific Products
Change view
Click to view available options
Includes:
13 Tuning Forks and One Case
Catalog No. Includes
S81831 13 Tuning Forks and One Case
1 options
This item is not returnable. View return policy
Catalog No. S81831 Supplier Supertek™ Supplier No. PH36128C/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Supertek™ Economy Chromatic Set of 13 Tuning Forks have been selected to illustrate the 12 tone 'Well Tempered' musical scale. These economy grade tuning forks are individually tested to assure that the proper pitch is achieved. Attractive and practical foam lined wooden storage case.
  • Includes 1 each of:
    • Middle C; 261.53 Hz
    • C#; 277.18 Hz
    • D; 293.66 Hz
    • D#; 311.13 Hz
    • E; 329.63 Hz
    • F; 349.28 Hz
    • F#; 369.99 Hz
    • G; 392 Hz
    • G#; 415.3 Hz
    • A; 440Hz
    • A#; 466.16 Hz
    • B; 493.88 Hz
    • C; 523.6 Hz
  • Comes in an attractive and practical foam lined wooden storage case.
Specifications
Product Type Tuning Fork
Includes 13 Tuning Forks and One Case
Frequency 261.53, 277.18, 293.66, 311.13, 329.63, 349.28, 369.99, 392, 415.3, 440, 466.16, 493.88, 523 Hz