Learn More
Omnimed Inc Economy Double Door Narcotic Cabinet (24”H x 16”W x 8”D)
SDP
Catalog No. NC3956395
Encompass_Preferred
Prevent unauthorized Personnel or patients from obtaining access to medications. Perfect for areas short on space or for mobile transit of medications our cabinet is made of heavy gauge steel with a beige powder coated finish and are built to meet schedule 2 drug storage requirements. They have strong piano hinged doors and a two door, dual flat key locking system to ensure content security. The outside and inner door are secured with a heavy gauge locking cam. Three extra keys are included for each lock. The ambi top of the cabinet lifts so that the unit can be flipped to accommodate left or right outer door access. Its two aluminum shelves are adjustable and increase storage capacity. Pre-drilled ensure stable wall attachment. Cabinets are are warranted for 5 Years with 1 year on the locks.
Catalog No. NC3956395 Supplier Omnimed Inc Supplier No. 182175
Prevent unauthorized Personnel or patients from obtaining access to medications. Perfect for areas short on space or for mobile transit of medications our cabinet is made of heavy gauge steel with a beige powder coated finish and are built to meet schedule 2 drug storage requirements. They have strong piano hinged doors and a two door, dual flat key locking system to ensure content security. The outside and inner door are secured with a heavy gauge locking cam. Three extra keys are included for each lock. The ambi top of the cabinet lifts so that the unit can be flipped to accommodate left or right outer door access. Its two aluminum shelves are adjustable and increase storage capacity. Pre-drilled ensure stable wall attachment. Cabinets are are warranted for 5 Years with 1 year on the locks.