Learn More
Grainger Techspray Aerosol Duster, Economy, HFC-152a, 10 oz.
Catalog No. 502793865
Encompass_Preferred
This economy duster is a 284 g (10 oz.) aerosol gas formulated with HFC-152a, offering an economical solution for cleaning delicate equipment. The moisture-free and residue-free duster provides a high-velocity gas stream to remove dust and debris from sensitive surfaces and electronic components. Note: Do not use on powered electrical equipment, hot surfaces, or around open flames and sparks.
  • Contains non-ozone depleting HFC-152a
  • Features lower global warming potential compared to HFC-134a
  • Safe for use on most plastics
  • Provides zero residue and moisture-free cleaning
  • VOC exempt according to EPA and CARB standards
  • Suitable for computers, printers, laboratory instruments, and circuit boards
  • Includes 60-month shelf life from date of manufacture
Catalog No. 50-279-3865 Supplier Grainger Supplier No. 19YY59
This economy duster is a 284 g (10 oz.) aerosol gas formulated with HFC-152a, offering an economical solution for cleaning delicate equipment. The moisture-free and residue-free duster provides a high-velocity gas stream to remove dust and debris from sensitive surfaces and electronic components. Note: Do not use on powered electrical equipment, hot surfaces, or around open flames and sparks.
  • Contains non-ozone depleting HFC-152a
  • Features lower global warming potential compared to HFC-134a
  • Safe for use on most plastics
  • Provides zero residue and moisture-free cleaning
  • VOC exempt according to EPA and CARB standards
  • Suitable for computers, printers, laboratory instruments, and circuit boards
  • Includes 60-month shelf life from date of manufacture