Learn More
MSC OccuNomix High Visibility Vest, Flame Resistant, Orange, X-Large, 1 ea
Catalog No. 5000686384
Encompass
This high visibility vest is designed for general purpose use where flame resistance is required. Constructed from a modacrylic and aramid blend, this solid orange vest provides protection while maintaining safety standards compliance.
  • Flame resistant material construction
  • ANSI 107-2015 compliant
  • ASTM F1506 certified
  • ATPV rating of 5.4 cal/cm²
  • NFPA 70E/HRC level 1 rating
  • Convenient hook and loop closure
  • Features 1 pocket for storage
  • Includes silver reflective stripes
Catalog No. 50-006-86384 Supplier MSC Supplier No. 69694578
This high visibility vest is designed for general purpose use where flame resistance is required. Constructed from a modacrylic and aramid blend, this solid orange vest provides protection while maintaining safety standards compliance.
  • Flame resistant material construction
  • ANSI 107-2015 compliant
  • ASTM F1506 certified
  • ATPV rating of 5.4 cal/cm²
  • NFPA 70E/HRC level 1 rating
  • Convenient hook and loop closure
  • Features 1 pocket for storage
  • Includes silver reflective stripes