Learn More
Grainger Economy Floor Mount Base Plate Triple
Catalog No. 502763866
Encompass_Preferred
The 80/20 Floor Mount 15 Series 16U359 is a floor-mounted aluminum frame designed for industrial applications. It has a load capacity of 250 lbs per shelf with 4 adjustable height shelves. The frame is made from T-slotted aluminum extrusion and measures 24"D x 36"W x 84"H. Assembly hardware is included. This sturdy, modular frame allows flexible configuration options for industrial and warehouse settings.
Catalog No. 50-276-3866 Supplier Grainger Supplier No. 16U359
The 80/20 Floor Mount 15 Series 16U359 is a floor-mounted aluminum frame designed for industrial applications. It has a load capacity of 250 lbs per shelf with 4 adjustable height shelves. The frame is made from T-slotted aluminum extrusion and measures 24"D x 36"W x 84"H. Assembly hardware is included. This sturdy, modular frame allows flexible configuration options for industrial and warehouse settings.