Learn More
Chemglass Life Sciences Economy Fluid Aspirator System
SDP
Catalog No. 501311393
Encompass_Preferred
Economy Fluid Aspirator System. Perform vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes. To use, simply insert a pipette tip, Pasteur pipette, or serological pipette into the collection tube and plug in to a 110/120V outlet. Self-contained system with bottle, tubing and vacuum pump is lightweight, compact and easy to use.Features: Built-in pump quickly aspirates fluids while maintaining a low noise level (76db at 1 meter) and minimal vibration Ideal for fluid collection from tissue and/or cell culture, Petri dishes, plates, flasks and tubes Chemical and corrosion resistant polypropylene collection bottle holds 3800mL (1 gallon) fluid wastes and has a safety release vent to prevent vacuum implosion A replaceable hydrophobic filter seals itself if it becomes wet protecting the pump motor from fluid contamination Compact design will not crowd workspace whether working in a hood or at the bench
Catalog No. 50-131-1393 Supplier Chemglass Life Sciences Supplier No. CLS910100
Add to Cart
Add to Cart
Economy Fluid Aspirator System. Perform vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes. To use, simply insert a pipette tip, Pasteur pipette, or serological pipette into the collection tube and plug in to a 110/120V outlet. Self-contained system with bottle, tubing and vacuum pump is lightweight, compact and easy to use.Features: Built-in pump quickly aspirates fluids while maintaining a low noise level (76db at 1 meter) and minimal vibration Ideal for fluid collection from tissue and/or cell culture, Petri dishes, plates, flasks and tubes Chemical and corrosion resistant polypropylene collection bottle holds 3800mL (1 gallon) fluid wastes and has a safety release vent to prevent vacuum implosion A replaceable hydrophobic filter seals itself if it becomes wet protecting the pump motor from fluid contamination Compact design will not crowd workspace whether working in a hood or at the bench