Learn More
MCR Safety Economy Goggle 23 Series
Catalog No. 19812778 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Clear lens
Catalog No.
19-812-778 Clear lens
1 options
This item is not returnable. View return policy
Catalog No. 19-812-778 Supplier MCR Safety Supplier No. 2310AF
This item is not returnable. View return policy
MCR Safety Economy Goggle 23 Series has a modern, stylish look, Duramass™ anti-fog and anti-scratch coatings and indirect ventilation.
Easy-to-adjust strap design allows each user a personal fit. Meets or exceeds ANSI Z87+ and CSA Z94.3.
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Frame Color Smoke
Ventilation Details Indirect vent
Frame Material PVC