Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Economy Hospital Body Bag with Envelope Zipper
SDP

Catalog No. 22444351 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
90 x 36in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-444-351 90 x 36in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-444-351 Supplier Mopec Supplier No. BE082

missing translation for 'validMainframeMsg'

Mopec Economy Hospital Body Bag with Envelope Zipper features a rust resistant nylon zipper with two pulls

  • No handles
  • Shelf life:10 years

Specifications

Closure Type Zip Closure
Color White
Material Virgin PVC
Quantity 1
Dimensions (L x W) 90 x 36in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.