Learn More
Harrick Plasma Basic Oil-Based Vacuum Pump 115V
SDP
Catalog No. NC9915936
Encompass_Preferred
The PDC-VPE is a rotary vane pump that uses hydrocarbon oil, suitable for pumping air or inert gases (N2, Ar) but is NOT compatible with pumping pure or concentrated oxygen (see our Oxygen Service Pumps). Pumping Speed (50/60 Hz): 5.1 m3/hr. Ultimate Total Pressure (gas ballast closed): 7.5 mTorr; Ultimate Total Pressure (gas ballast open): 150 mTorr; Includes check valve, hose, hose clamps, and hose adaptor to connect plasma cleaner to pump inlet. Includes oil mist filter at the pump outlet to filter oil vapor from pump exhaust; Size: 9.75″ H x 14″ W x 5.5″ D; Weight: 27 lb.
This item is not returnable. View return policy
Catalog No. NC9915936 Supplier Harrick Plasma Supplier No. PDCVPE
The PDC-VPE is a rotary vane pump that uses hydrocarbon oil, suitable for pumping air or inert gases (N2, Ar) but is NOT compatible with pumping pure or concentrated oxygen (see our Oxygen Service Pumps). Pumping Speed (50/60 Hz): 5.1 m3/hr. Ultimate Total Pressure (gas ballast closed): 7.5 mTorr; Ultimate Total Pressure (gas ballast open): 150 mTorr; Includes check valve, hose, hose clamps, and hose adaptor to connect plasma cleaner to pump inlet. Includes oil mist filter at the pump outlet to filter oil vapor from pump exhaust; Size: 9.75″ H x 14″ W x 5.5″ D; Weight: 27 lb.