Learn More
Harrick Plasma Economy Dry Oxygen Service Pump 115V
SDP
Catalog No. NC0139115
Encompass
The PDC-OPE pump uses no oil or fluid and is suitable for pumping oxygen, air, and inert gases (N2, Ar). The absence of pump oil eliminates the risk of oil backstreaming or contamination into the plasma chamber. Pumping speed (50/60 Hz): 3.0/3.6 m3/hr; Ultimate total pressure: 250 mTorr manufacturer’s specifications (in our lab tests, ultimate pressures of <100 mTorr were attained); Includes hose, hose clamps, hose adaptor, centering ring and swing clamp to connect plasma cleaner to pump inlet; Dimensions: 7″ H x 14″ W x 5.5″ D; Weight: 21 lb.
Catalog No. NC0139115 Supplier Harrick Plasma Supplier No. PDCOPE
The PDC-OPE pump uses no oil or fluid and is suitable for pumping oxygen, air, and inert gases (N2, Ar). The absence of pump oil eliminates the risk of oil backstreaming or contamination into the plasma chamber. Pumping speed (50/60 Hz): 3.0/3.6 m3/hr; Ultimate total pressure: 250 mTorr manufacturer’s specifications (in our lab tests, ultimate pressures of <100 mTorr were attained); Includes hose, hose clamps, hose adaptor, centering ring and swing clamp to connect plasma cleaner to pump inlet; Dimensions: 7″ H x 14″ W x 5.5″ D; Weight: 21 lb.