Learn More
Alkali Scientific Economy Screw cap Microtubes, 1.5 mL, Non-Sterile with O-ring, caps assembled, 500/ Pack
SDP
Catalog No. 502388860
Encompass_Preferred
2.0mL screw cap microcentrifuge tubes. Made from ultra-clear polypropylene, with crisp printed graduations, screw-cap tubes offer the best visualization of sample. May be centrifuged at up to 22,000xg. (standard non-graduated 1.5mL screw cap tubes are also available)
Catalog No. 50-238-8860 Supplier Alkali Scientific Supplier No. MT2210
2.0mL screw cap microcentrifuge tubes. Made from ultra-clear polypropylene, with crisp printed graduations, screw-cap tubes offer the best visualization of sample. May be centrifuged at up to 22,000xg. (standard non-graduated 1.5mL screw cap tubes are also available)