Learn More
World Precision Instrument Economy Tweezers #7, 11cm, Curved, 0.4x0.5mm Tips, packs of 6
SDP
Catalog No. NC9676713
Encompass
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.
Catalog No. NC9676713 Supplier World Precision Instrument Supplier No. 5019816
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.