Learn More
World Precision Instrument Economy Tweezers #1, 11cm, 0.45x0.55mm Tips
SDP
Catalog No. 502521369
Encompass
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.
Catalog No. 50-252-1369 Supplier World Precision Instrument Supplier No. 501974
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.