Learn More
World Precision Instrument Economy Tweezers #2, 11cm, 0.4x0.55mm Tips, packs of 6
SDP
Catalog No. 502539549
Encompass
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.
Catalog No. 50-253-9549 Supplier World Precision Instrument Supplier No. 5019756
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.