Learn More
World Precision Instrument Economy Tweezers #3, 11cm, 0.2x0.4mm Tips, Package of 6
SDP
Catalog No. 502539548
Encompass
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.
Catalog No. 50-253-9548 Supplier World Precision Instrument Supplier No. 5019766
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.