Learn More
World Precision Instrument Economy Tweezers #4, 11cm, 0.4x0.45mm Tips, pack of 6
SDP
Catalog No. 502539546
Encompass
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.
Catalog No. 50-253-9546 Supplier World Precision Instrument Supplier No. 5019786
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.