Learn More
World Precision Instrument Economy Tweezers #6, 11cm, 0.4x0.5mm Tips
SDP
Catalog No. 502521367
Encompass
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.
Catalog No. 50-252-1367 Supplier World Precision Instrument Supplier No. 501980
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.