Learn More
Electron Microscopy Sciences EMS Premium Economy Tweezers Style MM, S, 125 mm OAL
SDP
Catalog No. 501930954
Encompass_Preferred
The tweezers are made from 316 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 125 mm
OAL: Straight, thick, strong, pointed
Catalog No. 50-193-0954 Supplier Electron Microscopy Sciences Supplier No. 7832688
The tweezers are made from 316 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 125 mm
OAL: Straight, thick, strong, pointed