Learn More
Electron Microscopy Sciences EMS Premium Economy Tweezers Style SS, 140 mm OAL
SDP
Catalog No. 501930952
Encompass_Preferred
The tweezers are made from 316 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 140 mm
OAL: Straight, fine, sharp
Catalog No. 50-193-0952 Supplier Electron Microscopy Sciences Supplier No. 7832686
The tweezers are made from 316 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 140 mm
OAL: Straight, fine, sharp