Description
- Affordable vinyl floor tape for everyday use.
- Easy to apply.
- Suitable for facility marking and Lean/5S programs.
- Meets OSHA and ANSI guidelines for marking hazards and aisleways.
- Industrial-grade vinyl for light-to-medium traffic areas.
- Rubber-based adhesive adheres to most clean, dry surfaces.
Specifications
Specifications
| Product Type | Floor Marking Tape |
| Color | Black |
| Material | PVC |
| Adhesive Type | Rubber |