Learn More
Bio Rad Laboratories ECONOPAC 10DG DESALTING COLMN
Catalog No. NC9254907
Encompass
Econo-Pac 10DG Desalting Columns, 30pk, 10mL, Includes an upper frit, a snap-off end tip, and graduated column markings
Catalog No. NC9254907 Supplier Bio Rad Laboratories Supplier No. 7322010
Econo-Pac 10DG Desalting Columns, 30pk, 10mL, Includes an upper frit, a snap-off end tip, and graduated column markings