Learn More
Epoch Life Science Inc 96Well Batch DNA Binding Plate
Supplier: Epoch Life Science Inc 2020001
96-well filter plate with long drip for easier sample collection.850 ul volume capacity and 20-30 ug DNA binding capacity for each well. Compatible with most liquid handlers for automation.Requires centrifuge with swinging bucket rotors.Collection plates sold separately.