Learn More
Sklar Instruments Econo(TM) Sterile Knowles Bandage Scissors, Sklar , 5-1/2 in., Blunt/Blunt, Angled, Smooth, Stainless Steel, Disposable, Sterile. CS/50.
SDP
Catalog No. NC0964094 Shop All Sklar Instruments Products
Encompass_Preferred
Econo(TM) Sterile Knowles Bandage Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Angled, Smooth, Stainless Steel, Disposable, Sterile. Econo(TM) Sterile Knowles Bandage Scissors are durable, lightweight, and designed for cutting thick dressing material. Bandage scissors have a blunt tip on the upper blade so as not to cause any injury to the patient while removing bandages or dressings. Econo(TM) Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan. Econo(TM) Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo(TM) Sterile instruments significantly reduce the likelihood of cross-contamination. BX50.
Catalog No. NC0964094 Supplier Sklar Instruments Supplier No. 962402
Econo(TM) Sterile Knowles Bandage Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Angled, Smooth, Stainless Steel, Disposable, Sterile. Econo(TM) Sterile Knowles Bandage Scissors are durable, lightweight, and designed for cutting thick dressing material. Bandage scissors have a blunt tip on the upper blade so as not to cause any injury to the patient while removing bandages or dressings. Econo(TM) Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan. Econo(TM) Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo(TM) Sterile instruments significantly reduce the likelihood of cross-contamination. BX50.