Learn More
LGC GENOMICS LLC (Lucigen) EconoTaq DNA Polymerase (with Mg++), 1,000 UDependable Taq polymerase performance.Choice of reaction buffers, with or without MgCl2.Non-proofreading Polymerase
Catalog No. NC2160332
Encompass
EconoTaq DNA Polymerase is provided with a choice of 10X Reaction Buffer with Mg++ (with Mg++); or 10X Reaction Buffer without Mg++ and a separate tube of 25 mM MgCl2 (separate Mg++).
This item is not returnable. View return policy
Catalog No. NC2160332 Supplier LGC GENOMICS LLC Supplier No. 300311
EconoTaq DNA Polymerase is provided with a choice of 10X Reaction Buffer with Mg++ (with Mg++); or 10X Reaction Buffer without Mg++ and a separate tube of 25 mM MgCl2 (separate Mg++).