Learn More
Epoch Life Science Inc EconoSpin® RNA Mini Spin Columns w/ lid (50 columns with collection tubes)
SDP
Catalog No. NC1213037
Encompass_Preferred
Silica membrane mini spin columns with lid and collection tubes for RNA purification. Sample volume: 850 ul. Binding capacity: 45 ug RNA. Minimum elution volume: 20 ul. Compatible with third party buffer sets. 18 month shelf life. Used by labs around the world.
Catalog No. NC1213037 Supplier Epoch Life Science Inc Supplier No. 1940050
Silica membrane mini spin columns with lid and collection tubes for RNA purification. Sample volume: 850 ul. Binding capacity: 45 ug RNA. Minimum elution volume: 20 ul. Compatible with third party buffer sets. 18 month shelf life. Used by labs around the world.