Learn More
LGC GENOMICS LLC (Lucigen) EconoTaq PLUS GREEN 2X Master Mix, 500 RxnsDependable Taq polymerase performance.Choice of reaction buffers, with or without MgCl2.Non-proofreading Polymerase
Catalog No. NC0742407
Encompass
EconoTaq PLUS GREEN & EconoTaq PLUS 2X Master Mixes offer Performance, Convenience, Reliability, and Value for Routine PCR. More effective than other Taq master mixes. EconoTaq PLUS and PLUS GREEN 2X Master Mixes contain a proprietary PCR enhancer, so they can successfully amplify templates that fail to amplify with other PCR master mixes. High density reaction buffer enables direct gel loading following PCR. Excellent results in routine PCR. EconoTaq PLUS GREEN & EconoTaq PLUS contain high purity, high activity EconoTaq DNA Polymerase for reliable amplification of templates up to 5 kb. Amplify challenging GC-rich templates. These 2X Master Mixes can be cycled up to 98C to amplify challenging GC-rich templates others cannot. Convenient tracking dyes in EconoTaq PLUS GREEN. On a 1% agarose gel, the blue dye migrates at the same rate as a 5 kb DNA fragment, and the yellow dye migrates at 75 bp.
This item is not returnable. View return policy
Catalog No. NC0742407 Supplier LGC GENOMICS LLC Supplier No. 300331
EconoTaq PLUS GREEN & EconoTaq PLUS 2X Master Mixes offer Performance, Convenience, Reliability, and Value for Routine PCR. More effective than other Taq master mixes. EconoTaq PLUS and PLUS GREEN 2X Master Mixes contain a proprietary PCR enhancer, so they can successfully amplify templates that fail to amplify with other PCR master mixes. High density reaction buffer enables direct gel loading following PCR. Excellent results in routine PCR. EconoTaq PLUS GREEN & EconoTaq PLUS contain high purity, high activity EconoTaq DNA Polymerase for reliable amplification of templates up to 5 kb. Amplify challenging GC-rich templates. These 2X Master Mixes can be cycled up to 98C to amplify challenging GC-rich templates others cannot. Convenient tracking dyes in EconoTaq PLUS GREEN. On a 1% agarose gel, the blue dye migrates at the same rate as a 5 kb DNA fragment, and the yellow dye migrates at 75 bp.