Learn More
New England Biolabs, Inc. EcoO109I – 2000 units
SDP
Catalog No. 50812144
Encompass
A restriction endonuclease that recognizes the sequence RG^GNC_CY.
Catalog No. 50-812-144 Supplier New England Biolabs, Inc. Supplier No. R0503S
A restriction endonuclease that recognizes the sequence RG^GNC_CY.