Learn More
New England Biolabs, Inc. EcoP15I – 500 units
SDP
Catalog No. 50995268
Encompass
A restriction endonuclease that recognizes the sequence CAGCAGNNNNNNNNNNNNNNNNNNNNNNNNN^NN_.
Catalog No. 50-995-268 Supplier New England Biolabs, Inc. Supplier No. R0646S
A restriction endonuclease that recognizes the sequence CAGCAGNNNNNNNNNNNNNNNNNNNNNNNNN^NN_.