Learn More
Medchemexpress LLC Ecopipam hydrochloride | 190133-94-9 | 99.5% | 350.28 | 50 MG
SDP
Catalog No. 5000363043
Encompass_Preferred
Ecopipam hydrochloride (SCH 39166) is a potent, selective, and orally active antagonist of dopamine D1/D5 receptors, with Ki values of 1.2 nM and 2.0 nM, respectively. It demonstrates over 40-fold selectivity for D1/D5 receptors compared to D2, D4, 5-HT, and α2a receptors. This compound is suitable for research into conditions such as schizophrenia and obesity.
  • Potent and selective dopamine D1/D5 receptor antagonist
  • Exhibits high selectivity over D2, D4, 5-HT, and α2a receptors
  • Orally active compound
  • Utilized in schizophrenia and obesity research
  • Abolishes nicotine-induced enhancement of sensory reinforcers in adult rats
  • Antagonizes apomorphine-induced stereotypy in rats
Catalog No. 50-003-63043 Supplier Medchemexpress LLC Supplier No. HY1468950MG
Ecopipam hydrochloride (SCH 39166) is a potent, selective, and orally active antagonist of dopamine D1/D5 receptors, with Ki values of 1.2 nM and 2.0 nM, respectively. It demonstrates over 40-fold selectivity for D1/D5 receptors compared to D2, D4, 5-HT, and α2a receptors. This compound is suitable for research into conditions such as schizophrenia and obesity.
  • Potent and selective dopamine D1/D5 receptor antagonist
  • Exhibits high selectivity over D2, D4, 5-HT, and α2a receptors
  • Orally active compound
  • Utilized in schizophrenia and obesity research
  • Abolishes nicotine-induced enhancement of sensory reinforcers in adult rats
  • Antagonizes apomorphine-induced stereotypy in rats