Learn More
Medchemexpress LLC Ecopipam hydrochloride | 190133-94-9 | 99.5% | 350.28 | C19H21Cl2NO | 5 MG
SDP
Supplier:  Medchemexpress LLC HY146895MG
Encompass_Preferred
Ecopipam hydrochloride is a potent, selective, orally active antagonist of dopamine D1 and D5 receptors used in biochemical and pharmacological research. It has nanomolar affinity and shows high selectivity versus other monoaminergic receptors, and is supplied in high-purity forms suitable for in vitro and in vivo studies.
  • Potent, selective antagonist of dopamine D1 and D5 receptors.
  • Nanomolar affinity supports sensitive pharmacological assays.
  • Greater than 40-fold selectivity versus D2, D4, 5-HT, and α2a receptors.
  • Suitable for preclinical studies of neuropsychiatric and metabolic disorders.
  • Available as solid and as DMSO solution; provided at high purity.
Catalog No. 50-002-22988