Learn More
Medchemexpress LLC Ecopipam hydrochloride | 190133-94-9 | 99.5% | C19H21Cl2NO | 10MG
SDP
Catalog No. 5000258476
Encompass_Preferred
Ecopipam hydrochloride is the hydrochloride salt of ecopipam, a selective and orally active dopamine D1/D5 receptor antagonist used in research on dopaminergic signaling. It is supplied as a white to off-white solid with high reported purity and is intended for laboratory research applications.
  • Selective antagonist of dopamine D1/D5 receptors.
  • High purity suitable for research applications.
  • White to off-white solid, stable under recommended storage.
  • Molecular formula C19H21Cl2NO; molecular weight 350.28.
  • Packaged in small research quantities for laboratory studies.
Catalog No. 50-002-58476 Supplier Medchemexpress LLC Supplier No. HY1468910MG
Ecopipam hydrochloride is the hydrochloride salt of ecopipam, a selective and orally active dopamine D1/D5 receptor antagonist used in research on dopaminergic signaling. It is supplied as a white to off-white solid with high reported purity and is intended for laboratory research applications.
  • Selective antagonist of dopamine D1/D5 receptors.
  • High purity suitable for research applications.
  • White to off-white solid, stable under recommended storage.
  • Molecular formula C19H21Cl2NO; molecular weight 350.28.
  • Packaged in small research quantities for laboratory studies.