Learn More
TARGETMOL CHEMICALS INC ECOPLADIB 5MG
SDP
Catalog No. 502581562
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ecopladib (PLA 725) is a sub-micromolar inhibitor of cytosolic phospholipase A2(alpha) (cPLA2(alpha)) with IC50s of 0.15 uM and 0.11 uM in the GLU micelle and rat whole blood assays respectively. purity: 99%
Catalog No. 50-258-1562 Supplier TARGETMOL CHEMICALS INC Supplier No. T111495MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ecopladib (PLA 725) is a sub-micromolar inhibitor of cytosolic phospholipase A2(alpha) (cPLA2(alpha)) with IC50s of 0.15 uM and 0.11 uM in the GLU micelle and rat whole blood assays respectively. purity: 99%