Learn More
Justrite™ EcoPolyBlend™ Accumulation Center
GREENER_CHOICE
Catalog No. 19822314 Shop All Justrite Safety Group Products
Change view
Click to view available options
Holds:
6 Drums
8 Drums
Color:
Black
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Color
19-822-314 6 Drums Yellow
19-822-313 6 Drums Black
19-822-316 8 Drums Yellow
19-822-315 8 Drums Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-822-314 Supplier Justrite™ Supplier No. 28658
Add to Cart
Add to Cart
Create custom systems to deliver a complete solution. The Justrite™ EcoPolyBlend™ Accumulation Center combines units to create customized spill-control systems, allowing for the perfect solution to specific industry needs.
Low-Profile EcoPolyBlend™ Accumulation Centers only measure 5.5 in. (140-mm) high for easy handling of drums. Made of recycled polyethylene, these accumulation centers are nearly impervious to chemicals, including acids and corrosives. Seamless construction eliminates leaks, making these ideal for drum filling and dispensing operations. Self-locating grates won’t ride-up, and are removable for easy cleaning. Combine 2-, 4-, 6-, and 8-drum modular units to create your own customized system. Use the optional joining clips to firmly lock units together. Sumps remain separate and clips can be removed for easy repositioning.
Specifications
Color Yellow
Holds 6 Drums