Learn More
Justrite™ EcoPolyBlend™ Accumulation Center
GSA_VAGREENER_CHOICE
Catalog No. 19822316 Shop All Justrite Safety Group Products
Change view
Click to view available options
Holds:
6 Drums
8 Drums
Color:
Black
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Color
19-822-316 8 Drums Yellow
19-822-314 6 Drums Yellow
19-822-313 6 Drums Black
19-822-315 8 Drums Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-822-316 Supplier Justrite™ Supplier No. 28660
Add to Cart
Add to Cart
Create custom systems to deliver a complete solution. The Justrite™ EcoPolyBlend™ Accumulation Center combines units to create customized spill-control systems, allowing for the perfect solution to specific industry needs.
Low-Profile EcoPolyBlend™ Accumulation Centers only measure 5.5 in. (140-mm) high for easy handling of drums. Made of recycled polyethylene, these accumulation centers are nearly impervious to chemicals, including acids and corrosives. Seamless construction eliminates leaks, making these ideal for drum filling and dispensing operations. Self-locating grates won’t ride-up, and are removable for easy cleaning. Combine 2-, 4-, 6-, and 8-drum modular units to create your own customized system. Use the optional joining clips to firmly lock units together. Sumps remain separate and clips can be removed for easy repositioning.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Color Yellow
Holds 8 Drums