Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ EcoPolyBlend™ Spill Control Pallets
GREENER_CHOICESCI_ED

Catalog No. S28764 Shop All Justrite Safety Group Products
Change view
Click to view available options
Includes:
With drain
Without drain
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S28764 Without drain
S28765 With drain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S28764 Supplier Justrite™ Supplier No. 28623
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Keep workers safe from accidental slips indoors and avoid costly hazmat cleanup costs. Justrite™ EcoPolyBlend™ Spill Control Pallets can withstand the toughest environments for superior spill protection. Black pallets are constructed of 100% recycled content and the dark color camouflages dirt and keeps work places looking clean.

  • Protect against groundwater contamination, using post-industrial material
  • Robust pallets withstand the toughest environments for superior spill control
  • Durable, UV-protected polyethylene is nearly impervious to chemical attack from hazardous acids, caustics, and solvents
  • Seamless construction eliminates leaks—even if a 55-gallon (208L) drum fails and completely empties
  • Oversized leakproof sumps meet or exceed regulations
  • Sturdy, removable 2.5 in. (64mm) thick grates offer superior crack resistance; self-locating
  • Black pallets constructed of 100% recycled content; dark color camouflages dirt and keeps work places looking clean
  • Yellow pallets ideal for high visibility or liquid segregation; virgin poly sumps with 100% recycled grates (a total of 35% recycled content)
  • Square pallet may be equipped with an optional ramp
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail

Specifications

Certifications/Compliance SPCC, EPA, NFPA, U (NFPA), meets fire codes
Color Black
Capacity (English) 66 gal.
Capacity (Metric) 250 L
Material Polyethylene
Type Ecopolyblend Spill Control Pallet
For Use With (Application) Keep workers safe from accidental slips indoors and avoid costly hazmat cleanup costs, protect against groundwater contamination
For Use With (Equipment) Drums
Holds 2 Drums
Includes Without drain
Load Bearing Capacity (English) 2500 lb.
Load Bearing Capacity (Metric) 1134 kg
Resistant Chemical
Width (English) 49 in.
Width (Metric) 124.5 cm
Height (English) 18 in.
Height (Metric) 45.7 cm
Depth (English) 25 in.
Depth (Metric) 63.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.