Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ EcoPolyBlend™ Spill Tray
GREENER_CHOICE

Catalog No. 19822510 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
20 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-822-510 20 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-822-510 Supplier Justrite™ Supplier No. 28717
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Increase the efficiency and space of existing cabinetry. The Justrite™ EcoPolyBlend™ Spill Tray is a lightweight and low-profile design for small-scale spill control.

Made of 100% recycled polyethylene, these eco-friendly spill trays will not rust or corrode. Raised rib bottom adds rigidity and keeps contents out of spills—perfect for pouring and filling applications. Rated for service temperatures between –30° to +120°F (–34° to +49°C).

Specifications

Color Black
Capacity (English) 20 gal.
Capacity (Metric) 76 L
Material Polyethylene
For Use With (Application) Small-scale spill control
Dimensions (L x W x H) 584 x 1207 x 140mm (23 x 47.5 x 5.5 in.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.