Learn More
New England Biolabs, Inc. EcoRI – 10000 units
SDP
Catalog No. 50811920
Encompass
A restriction endonuclease that recognizes the sequence G^AATT_C.
Catalog No. 50-811-920 Supplier New England Biolabs, Inc. Supplier No. R0101S
A restriction endonuclease that recognizes the sequence G^AATT_C.