Learn More
New England Biolabs, Inc. EcoRI Methyltransferase – 10000 units
SDP
Catalog No. 50811625
Encompass
EcoRI Methyltransferase modifies the internal adenine residue (N6) of the sequence GAATTC.
Catalog No. 50-811-625 Supplier New England Biolabs, Inc. Supplier No. M0211S
EcoRI Methyltransferase modifies the internal adenine residue (N6) of the sequence GAATTC.