Learn More
New England Biolabs, Inc. EcoRV – 20000 units
SDP
Supplier:  New England Biolabs, Inc. R0195M
Encompass
A restriction endonuclease that recognizes the sequence GAT_^ATC.
Catalog No. 50-812-125