Learn More
New England Biolabs, Inc. EcoRV – 4000 units
SDP
Catalog No. 50812127
Encompass
A restriction endonuclease that recognizes the sequence GAT_^ATC.
Catalog No. 50-812-127 Supplier New England Biolabs, Inc. Supplier No. R0195T
A restriction endonuclease that recognizes the sequence GAT_^ATC.