Learn More
New England Biolabs, Inc. EcoRV – 20000 units
SDP
Catalog No. 50812124
Encompass
A restriction endonuclease that recognizes the sequence GAT_^ATC.
Catalog No. 50-812-124 Supplier New England Biolabs, Inc. Supplier No. R0195L
A restriction endonuclease that recognizes the sequence GAT_^ATC.