Learn More
New England Biolabs, Inc. EcoRV – 4000 units
SDP
Catalog No. 50812126
Encompass
A restriction endonuclease that recognizes the sequence GAT_^ATC.
Catalog No. 50-812-126 Supplier New England Biolabs, Inc. Supplier No. R0195S
A restriction endonuclease that recognizes the sequence GAT_^ATC.