Learn More
Lovibond™ EcoSafe Arsenic Test Kit
Catalog No. 09051517 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
50 mL
Catalog No. Volume (Metric)
09-051-517 50 mL
1 options
Catalog No. 09-051-517 Supplier Lovibond™ Supplier No. 475250
Add to Cart
Add to Cart
Designed for on-site use, results can be obtained in as little as 7 mins. Intuitive pictogram instructions and color-coded components ensure error-free operation.
  • Mercury-free innovation for safer testing
  • Fast, reliable field testing with user-friendly design
  • The sustainable choice for global water safety
  • Meets World Health Organization’s recommendation of max. 10 μg/L arsenic in drinking water by ensuring reliable detection below that level
  • Simple, fast, and accurate testing of arsenic from 2 to 500 μg/L
Specifications
pH Range 0 to 14
Volume (Metric) 50 mL
Type Arsenic Test Kit