Learn More
National Diagnostics ECOSCINT O 4X4LT/CS
SDP
Catalog No. 5089990188
Encompass
Ecoscint O, High efficiency, biodegradable scintillation cocktail for nonaqueous samples, 4X4LT/CS
Catalog No. 50-899-90188 Supplier National Diagnostics Supplier No. LS2744X4LTRCS
Ecoscint O, High efficiency, biodegradable scintillation cocktail for nonaqueous samples, 4X4LT/CS