Learn More
MSC Ecolab Metal Polish, Water-Based, Stainless Steel, 6 x 32 oz
Catalog No. 502836821
Encompass
This metal polish is a water-based solution designed for cleaning and polishing stainless steel surfaces. It comes ready to use and is free of solvents and butyl. The product is intended for industrial and professional use to maintain metal surfaces.
  • Water-based formula
  • Cleans and polishes stainless steel
  • Ready to use
  • Solvent and butyl-free
  • Supplied in 6-pack of 32 oz containers
  • Suitable for industrial and professional applications
Catalog No. 50-283-6821 Supplier MSC Supplier No. 59852970
Add to Cart
Add to Cart
This metal polish is a water-based solution designed for cleaning and polishing stainless steel surfaces. It comes ready to use and is free of solvents and butyl. The product is intended for industrial and professional use to maintain metal surfaces.
  • Water-based formula
  • Cleans and polishes stainless steel
  • Ready to use
  • Solvent and butyl-free
  • Supplied in 6-pack of 32 oz containers
  • Suitable for industrial and professional applications