Learn More
Medchemexpress LLC Ecotin E. coli His 100ug | 100ug
SDP
Catalog No. 5000194969
Encompass_Preferred
Ecotin a bacterial protein inhibitor regulates serine proteases like trypsin and subtilisin Ecotin s unique surface loop forms a stable complex influencing enzymatic activity Ecotin Protein E coli (His) is the recombinant E coli-derived Ecotin protein expressed by E coli with C-His labeled tag
Catalog No. 50-001-94969 Supplier Medchemexpress LLC Supplier No. HYP79083100UG
Ecotin a bacterial protein inhibitor regulates serine proteases like trypsin and subtilisin Ecotin s unique surface loop forms a stable complex influencing enzymatic activity Ecotin Protein E coli (His) is the recombinant E coli-derived Ecotin protein expressed by E coli with C-His labeled tag