Learn More
Research Products International Corp eCount Electronic Colony Counter
SDP
Catalog No. 501977314
Encompass
eCount combines a precision electronic counter with a Sharpie pen for marking petri dishes when counting colonies. Unique counter prevents counting errors such as double counting of colonies. Features easy to use touch controls and a visual or audible signal to identify each count. Includes on fine point black Sharpie pen and stand.
Package includes counter, black Sharpie pen, CR1225 3.0 Lithium Battery, hand tally adapter and weighted stand.
Catalog No. 50-197-7314 Supplier Research Products International Corp Supplier No. 810530
eCount combines a precision electronic counter with a Sharpie pen for marking petri dishes when counting colonies. Unique counter prevents counting errors such as double counting of colonies. Features easy to use touch controls and a visual or audible signal to identify each count. Includes on fine point black Sharpie pen and stand.
Package includes counter, black Sharpie pen, CR1225 3.0 Lithium Battery, hand tally adapter and weighted stand.