Learn More
Hach Company ECR Masking Reagent Solution, 25 mL SCDB
Supplier:  Hach Company 2380123
Encompass
For Aluminum determination by the Eriochrome Cyanine R Method. Hach method 8326. 25 mL Self-Contained Dropping Bottle
Catalog No. 50-199-5898