Learn More
Hach Company ECR Reagent Powder Pillows, 20 mL, pk/100
Supplier:  Hach Company 2603849
Encompass
For determination of Aluminum by the Eriochrome Cyanine R (ECR) Method. Hach Method 8326. Sample Size: 20 mL (split) Range: 0.002 - 0.250 mg/L Al. Pack of 100 Powder Pillows
Catalog No. 50-199-6319