Learn More
Hach Company ECR Reagent Powder Pillows, 50 mL, pk/25
Supplier:  Hach Company 2380268
Encompass
For Aluminum determination by the Eriochrome Cyanine R Method. Hach method 8326. Sample size: 25 mL, Pack of 25 Powder Pillows.
Catalog No. 50-199-5899